Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613250 MIM: 613822 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EBLN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
EBLN2 - endogenous Bornavirus-like nucleoprotein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018029.4 | 559 | Missense Mutation | CCC,TCC | P,S 46 | NP_060499.3 |
PPP4R2 - protein phosphatase 4 regulatory subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318025.1 | 559 | Intron | NP_001304954.1 | |||
NM_001318026.1 | 559 | Intron | NP_001304955.1 | |||
NM_001318027.1 | 559 | Intron | NP_001304956.1 | |||
NM_001318028.1 | 559 | Intron | NP_001304957.1 | |||
NM_174907.3 | 559 | Intron | NP_777567.1 |