Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTTTTTTGATTGTTTCCTGTGTT[C/T]TTTCTTCTGTTTCCCTTTTCCTGCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616475 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEP72 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CEP72 - centrosomal protein 72 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018140.3 | Intron | NP_060610.2 | ||||
XM_005248322.3 | Intron | XP_005248379.1 | ||||
XM_011514063.1 | Intron | XP_011512365.1 | ||||
XM_011514064.2 | Intron | XP_011512366.1 | ||||
XM_017009626.1 | Intron | XP_016865115.1 | ||||
XM_017009627.1 | Intron | XP_016865116.1 |
LOC100996325 - uncharacterized LOC100996325 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |