Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605008 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAMTS6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ADAMTS6 - ADAM metallopeptidase with thrombospondin type 1 motif 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_197941.3 | Intron | NP_922932.2 | ||||
XM_011543113.2 | Intron | XP_011541415.1 | ||||
XM_011543114.2 | Intron | XP_011541416.1 | ||||
XM_011543115.2 | Intron | XP_011541417.1 | ||||
XM_011543116.2 | Intron | XP_011541418.1 | ||||
XM_011543117.1 | Intron | XP_011541419.1 | ||||
XM_011543119.2 | Intron | XP_011541421.1 | ||||
XM_011543120.2 | Intron | XP_011541422.1 | ||||
XM_011543121.2 | Intron | XP_011541423.2 | ||||
XM_011543122.2 | Intron | XP_011541424.1 | ||||
XM_011543123.2 | Intron | XP_011541425.1 | ||||
XM_011543124.2 | Intron | XP_011541426.1 | ||||
XM_011543125.1 | Intron | XP_011541427.1 | ||||
XM_011543126.2 | Intron | XP_011541428.1 | ||||
XM_011543127.2 | Intron | XP_011541429.1 |
Set Membership: |
HapMap Validated |