Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTCATGTCAAAGGCTGGCGAGTC[A/G]TGGGCCTCAACACCCAGGTAGCGGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DUSP26 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DUSP26 - dual specificity phosphatase 26 (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305115.1 | 639 | Silent Mutation | CAC,CAT | H,H 119 | NP_001292044.1 | |
NM_001305116.1 | 639 | Silent Mutation | CAC,CAT | H,H 119 | NP_001292045.1 | |
NM_024025.2 | 639 | Silent Mutation | CAC,CAT | H,H 119 | NP_076930.1 |