Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ECT2L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ECT2L - epithelial cell transforming 2 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077706.2 | Intron | NP_001071174.1 | ||||
NM_001195037.2 | Intron | NP_001181966.1 | ||||
XM_006715472.3 | Intron | XP_006715535.1 | ||||
XM_011535795.2 | Intron | XP_011534097.1 | ||||
XM_011535797.2 | Intron | XP_011534099.1 | ||||
XM_017010828.1 | Intron | XP_016866317.1 | ||||
XM_017010829.1 | Intron | XP_016866318.1 | ||||
XM_017010830.1 | Intron | XP_016866319.1 |