Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613022 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
OGDH PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
OGDH - oxoglutarate dehydrogenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003941.2 | Intron | NP_001003941.1 | ||||
NM_001165036.1 | Intron | NP_001158508.1 | ||||
NM_002541.3 | Intron | NP_002532.2 | ||||
XM_005249759.4 | Intron | XP_005249816.1 | ||||
XM_005249761.3 | Intron | XP_005249818.1 | ||||
XM_011515408.2 | Intron | XP_011513710.1 |
Set Membership: |
HapMap |