Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 140572 MIM: 604548 MIM: 610788 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSP90AB1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
HSP90AB1 - heat shock protein 90 alpha family class B member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271969.1 | 1507 | Intron | NP_001258898.1 | |||
NM_001271970.1 | 1507 | Intron | NP_001258899.1 | |||
NM_001271971.1 | 1507 | Intron | NP_001258900.1 | |||
NM_001271972.1 | 1507 | Intron | NP_001258901.1 | |||
NM_007355.3 | 1507 | Intron | NP_031381.2 |
MIR4647 - microRNA 4647 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NFKBIE - NFKB inhibitor epsilon | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC35B2 - solute carrier family 35 member B2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286509.1 | 1507 | UTR 3 | NP_001273438.1 | |||
NM_001286510.1 | 1507 | UTR 3 | NP_001273439.1 | |||
NM_001286511.1 | 1507 | UTR 3 | NP_001273440.1 | |||
NM_001286512.1 | 1507 | UTR 3 | NP_001273441.1 | |||
NM_001286513.1 | 1507 | UTR 3 | NP_001273442.1 | |||
NM_001286517.1 | 1507 | UTR 3 | NP_001273446.1 | |||
NM_001286519.1 | 1507 | UTR 3 | NP_001273448.1 | |||
NM_001286520.1 | 1507 | UTR 3 | NP_001273449.1 | |||
NM_178148.3 | 1507 | UTR 3 | NP_835361.1 |
Set Membership: |
HapMap Validated |