Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612035 MIM: 186975 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AARS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AARS2 - alanyl-tRNA synthetase 2, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020745.3 | 3072 | Intron | NP_065796.1 | |||
XM_005249245.3 | 3072 | Intron | XP_005249302.1 | |||
XM_011514764.2 | 3072 | UTR 3 | XP_011513066.1 | |||
XM_017011112.1 | 3072 | Intron | XP_016866601.1 |
TCTE1 - t-complex-associated-testis-expressed 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182539.3 | 3072 | Intron | NP_872345.2 | |||
XM_005248872.1 | 3072 | Intron | XP_005248929.1 | |||
XM_005248873.2 | 3072 | Intron | XP_005248930.1 | |||
XM_005248874.2 | 3072 | Intron | XP_005248931.1 | |||
XM_006715009.3 | 3072 | Intron | XP_006715072.1 | |||
XM_011514336.2 | 3072 | Intron | XP_011512638.1 | |||
XM_011514338.2 | 3072 | Intron | XP_011512640.1 | |||
XM_011514339.2 | 3072 | Intron | XP_011512641.1 | |||
XM_011514340.1 | 3072 | Intron | XP_011512642.1 |