Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 173510 | ||||||||||||||||||||
Literature Links: |
CD36 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CD36 - CD36 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000072.3 | Intron | NP_000063.2 | ||||
NM_001001547.2 | Intron | NP_001001547.1 | ||||
NM_001001548.2 | Intron | NP_001001548.1 | ||||
NM_001127443.1 | Intron | NP_001120915.1 | ||||
NM_001127444.1 | Intron | NP_001120916.1 | ||||
NM_001289908.1 | Intron | NP_001276837.1 | ||||
NM_001289909.1 | Intron | NP_001276838.1 | ||||
NM_001289911.1 | Intron | NP_001276840.1 | ||||
XM_005250713.1 | Intron | XP_005250770.1 | ||||
XM_005250714.1 | Intron | XP_005250771.1 | ||||
XM_005250715.4 | Intron | XP_005250772.1 |