Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||
Literature Links: |
NCK1-AS1 PubMed Links | |||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS - Not Available | African American
|
YRI (Yoruba) - Not Available | ||||||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR - Not Available | ||||||||
AMR - Not Available |
NCK1-AS1 - NCK1 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC35G2 - solute carrier family 35 member G2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001097599.1 | Intron | NP_001091068.1 | ||||
NM_001097600.1 | Intron | NP_001091069.1 | ||||
NM_025246.2 | Intron | NP_079522.2 | ||||
XM_006713773.3 | Intron | XP_006713836.1 | ||||
XM_011513214.2 | Intron | XP_011511516.1 | ||||
XM_017007289.1 | Intron | XP_016862778.1 | ||||
XM_017007290.1 | Intron | XP_016862779.1 | ||||
XM_017007291.1 | Intron | XP_016862780.1 |
Set Membership: |
Validated |