Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607033 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLDN20 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CLDN20 - claudin 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001346.3 | Intron | NP_001001346.1 | ||||
XM_011535848.2 | Intron | XP_011534150.1 |
TFB1M - transcription factor B1, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016020.3 | Intron | NP_057104.2 | ||||
XM_005267006.4 | Intron | XP_005267063.1 | ||||
XM_011535870.1 | Intron | XP_011534172.1 | ||||
XM_011535871.1 | Intron | XP_011534173.1 | ||||
XM_011535872.2 | Intron | XP_011534174.1 | ||||
XM_011535873.2 | Intron | XP_011534175.1 | ||||
XM_017010917.1 | Intron | XP_016866406.1 |
Set Membership: |
HapMap Validated |