Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 125855 MIM: 155550 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DGKA PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DGKA - diacylglycerol kinase alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001345.4 | 1881 | Intron | NP_001336.2 | |||
NM_201444.2 | 1881 | Intron | NP_958852.1 | |||
NM_201445.1 | 1881 | Intron | NP_958853.1 | |||
NM_201554.1 | 1881 | Intron | NP_963848.1 | |||
XM_005268688.2 | 1881 | Intron | XP_005268745.1 | |||
XM_005268689.2 | 1881 | Intron | XP_005268746.1 | |||
XM_005268690.2 | 1881 | Intron | XP_005268747.1 | |||
XM_011537991.2 | 1881 | Intron | XP_011536293.1 | |||
XM_011537993.2 | 1881 | Intron | XP_011536295.1 | |||
XM_011537995.2 | 1881 | Intron | XP_011536297.1 | |||
XM_017018900.1 | 1881 | Intron | XP_016874389.1 | |||
XM_017018901.1 | 1881 | Intron | XP_016874390.1 | |||
XM_017018902.1 | 1881 | Intron | XP_016874391.1 | |||
XM_017018903.1 | 1881 | Intron | XP_016874392.1 | |||
XM_017018904.1 | 1881 | Intron | XP_016874393.1 | |||
XM_017018905.1 | 1881 | Intron | XP_016874394.1 | |||
XM_017018906.1 | 1881 | Intron | XP_016874395.1 | |||
XM_017018907.1 | 1881 | Intron | XP_016874396.1 | |||
XM_017018908.1 | 1881 | Intron | XP_016874397.1 | |||
XM_017018909.1 | 1881 | Intron | XP_016874398.1 |
PMEL - premelanosome protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001200053.1 | 1881 | Silent Mutation | AAC,AAT | N,N 566 | NP_001186982.1 | |
NM_001200054.1 | 1881 | Silent Mutation | AAC,AAT | N,N 659 | NP_001186983.1 | |
NM_001320121.1 | 1881 | Silent Mutation | AAC,AAT | N,N 617 | NP_001307050.1 | |
NM_001320122.1 | 1881 | Silent Mutation | AAC,AAT | N,N 610 | NP_001307051.1 | |
NM_006928.4 | 1881 | Silent Mutation | AAC,AAT | N,N 652 | NP_008859.1 | |
XM_006719569.1 | 1881 | Silent Mutation | AAC,AAT | N,N 652 | XP_006719632.1 | |
XM_011538685.1 | 1881 | Silent Mutation | AAC,AAT | N,N 659 | XP_011536987.1 |
Set Membership: |
HapMap |