Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612104 MIM: 608730 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD52 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ANKRD52 - ankyrin repeat domain 52 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NABP2 - nucleic acid binding protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF41 - ring finger protein 41 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC39A5 - solute carrier family 39 member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135195.1 | 175 | UTR 5 | NP_001128667.1 | |||
NM_173596.2 | 175 | UTR 5 | NP_775867.2 | |||
XM_005268803.1 | 175 | UTR 5 | XP_005268860.1 | |||
XM_011538198.1 | 175 | UTR 5 | XP_011536500.1 | |||
XM_011538199.1 | 175 | UTR 5 | XP_011536501.1 | |||
XM_011538200.1 | 175 | UTR 5 | XP_011536502.1 | |||
XM_011538201.1 | 175 | UTR 5 | XP_011536503.1 | |||
XM_017019185.1 | 175 | UTR 5 | XP_016874674.1 | |||
XM_017019186.1 | 175 | UTR 5 | XP_016874675.1 | |||
XM_017019187.1 | 175 | UTR 5 | XP_016874676.1 |
Set Membership: |
HapMap |