Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF41 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RNF41 - ring finger protein 41 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242826.1 | 3572 | UTR 3 | NP_001229755.1 | |||
NM_005785.3 | 3572 | UTR 3 | NP_005776.1 | |||
NM_194358.2 | 3572 | UTR 3 | NP_919339.1 | |||
NM_194359.2 | 3572 | UTR 3 | NP_919340.1 | |||
XM_005268561.4 | 3572 | UTR 3 | XP_005268618.1 | |||
XM_011537734.2 | 3572 | UTR 3 | XP_011536036.1 | |||
XM_011537735.2 | 3572 | UTR 3 | XP_011536037.1 |