Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604002 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PQLC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PQLC3 - PQ loop repeat containing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ROCK2 - Rho associated coiled-coil containing protein kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321643.1 | 5029 | Intron | NP_001308572.1 | |||
NM_004850.4 | 5029 | Intron | NP_004841.2 | |||
XM_005246190.3 | 5029 | Intron | XP_005246247.1 | |||
XM_011510417.2 | 5029 | Intron | XP_011508719.1 | |||
XM_017005378.1 | 5029 | UTR 3 | XP_016860867.1 | |||
XM_017005379.1 | 5029 | Intron | XP_016860868.1 |
Set Membership: |
HapMap |