Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602895 MIM: 608066 | ||||||||||||||||||||
Literature Links: |
SAFB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SAFB - scaffold attachment factor B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201338.1 | 217 | Missense Mutation | CCG,GCG | P,A 16 | NP_001188267.1 | |
NM_001201339.1 | 217 | Missense Mutation | CCG,GCG | P,A 16 | NP_001188268.1 | |
NM_001201340.1 | 217 | Missense Mutation | CCG,GCG | P,A 16 | NP_001188269.1 | |
NM_001320571.1 | 217 | UTR 5 | NP_001307500.1 | |||
NM_001320572.1 | 217 | Missense Mutation | CCG,GCG | P,A 16 | NP_001307501.1 | |
NM_002967.3 | 217 | Missense Mutation | CCG,GCG | P,A 16 | NP_002958.2 | |
XM_017027114.1 | 217 | UTR 5 | XP_016882603.1 | |||
XM_017027115.1 | 217 | UTR 5 | XP_016882604.1 | |||
XM_017027116.1 | 217 | UTR 5 | XP_016882605.1 | |||
XM_017027117.1 | 217 | UTR 5 | XP_016882606.1 | |||
XM_017027118.1 | 217 | UTR 5 | XP_016882607.1 |
SAFB2 - scaffold attachment factor B2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014649.2 | 217 | Intron | NP_055464.1 | |||
XM_011528449.2 | 217 | Intron | XP_011526751.1 | |||
XM_011528450.2 | 217 | Intron | XP_011526752.1 | |||
XM_017027520.1 | 217 | Intron | XP_016883009.1 | |||
XM_017027521.1 | 217 | Intron | XP_016883010.1 | |||
XM_017027522.1 | 217 | Intron | XP_016883011.1 |