Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600188 MIM: 601041 | ||||||||||||||||||||
Literature Links: |
AES PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AES - amino-terminal enhancer of split | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130.5 | 1266 | UTR 3 | NP_001121.2 | |||
NM_198969.1 | 1266 | UTR 3 | NP_945320.1 | |||
NM_198970.1 | 1266 | UTR 3 | NP_945321.1 | |||
XM_006722664.1 | 1266 | UTR 3 | XP_006722727.1 |
TLE2 - transducin like enhancer of split 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |