Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGTGTTCTGGAAGACTCCGTGGA[G/T]CGGGACGCAGGCACTGCTGTTTGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142765 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100128568 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LOC100128568 - uncharacterized LOC100128568 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RFX2 - regulatory factor X2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000635.3 | 3166 | UTR 3 | NP_000626.2 | |||
NM_134433.2 | 3166 | UTR 3 | NP_602309.1 | |||
XM_011528171.2 | 3166 | UTR 3 | XP_011526473.1 | |||
XM_011528172.1 | 3166 | UTR 3 | XP_011526474.1 | |||
XM_017027107.1 | 3166 | UTR 3 | XP_016882596.1 | |||
XM_017027108.1 | 3166 | UTR 3 | XP_016882597.1 |
Set Membership: |
HapMap Validated |