Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602565 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCL25 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CCL25 - C-C motif chemokine ligand 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201359.1 | 178 | Missense Mutation | ACC,GCC | T,A 23 | NP_001188288.1 | |
NM_005624.3 | 178 | Missense Mutation | ACC,GCC | T,A 23 | NP_005615.2 | |
XM_011528177.2 | 178 | Missense Mutation | ACC,GCC | T,A 23 | XP_011526479.1 | |
XM_017027119.1 | 178 | Missense Mutation | ACC,GCC | T,A 83 | XP_016882608.1 | |
XM_017027120.1 | 178 | Missense Mutation | ACC,GCC | T,A 23 | XP_016882609.1 | |
XM_017027121.1 | 178 | Missense Mutation | ACC,GCC | T,A 23 | XP_016882610.1 | |
XM_017027122.1 | 178 | Missense Mutation | ACC,GCC | T,A 23 | XP_016882611.1 |
Set Membership: |
HapMap |