Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604672 MIM: 616256 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD209 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CD209 - CD209 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144893.1 | 2101 | UTR 3 | NP_001138365.1 | |||
NM_001144894.1 | 2101 | UTR 3 | NP_001138366.1 | |||
NM_001144895.1 | 2101 | UTR 3 | NP_001138367.1 | |||
NM_001144896.1 | 2101 | UTR 3 | NP_001138368.1 | |||
NM_001144897.1 | 2101 | UTR 3 | NP_001138369.1 | |||
NM_001144899.1 | 2101 | UTR 3 | NP_001138371.1 | |||
NM_021155.3 | 2101 | UTR 3 | NP_066978.1 | |||
XM_005272472.4 | 2101 | UTR 3 | XP_005272529.1 |
CLEC4G - C-type lectin domain family 4 member G | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |