Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
41 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 186780 MIM: 164175 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD247 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CD247 - CD247 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000734.3 | 1551 | UTR 3 | NP_000725.1 | |||
NM_198053.2 | 1551 | UTR 3 | NP_932170.1 | |||
XM_011510144.2 | 1551 | UTR 3 | XP_011508446.1 | |||
XM_011510145.1 | 1551 | UTR 3 | XP_011508447.1 | |||
XM_017002800.1 | 1551 | UTR 3 | XP_016858289.1 | |||
XM_017002801.1 | 1551 | UTR 3 | XP_016858290.1 |
POU2F1 - POU class 2 homeobox 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap JSNP |