Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTCTTTTTATTTTACTAAATTTT[A/G]TGGTTGTGAACCGAAGTAATATATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
40 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610134 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ST6GALNAC5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ST6GALNAC5 - ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320273.1 | Intron | NP_001307202.1 | ||||
NM_001320274.1 | Intron | NP_001307203.1 | ||||
NM_030965.2 | Intron | NP_112227.1 |