Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 615125 MIM: 602010 | ||||||||||||||||||||
Literature Links: |
ANKRD13C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANKRD13C - ankyrin repeat domain 13C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030816.4 | 2435 | UTR 3 | NP_110443.3 | |||
XM_005271234.3 | 2435 | UTR 3 | XP_005271291.1 | |||
XM_005271235.3 | 2435 | UTR 3 | XP_005271292.1 | |||
XM_006710929.3 | 2435 | UTR 3 | XP_006710992.1 | |||
XM_017002413.1 | 2435 | UTR 3 | XP_016857902.1 | |||
XM_017002414.1 | 2435 | UTR 3 | XP_016857903.1 | |||
XM_017002415.1 | 2435 | UTR 3 | XP_016857904.1 | |||
XM_017002416.1 | 2435 | UTR 3 | XP_016857905.1 |
SRSF11 - serine and arginine rich splicing factor 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |