Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTAGTCACCTAACACCAAGTGTCC[C/G]TGATACATAAGAATTGATTTGCAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
39 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603609 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FPGT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FPGT - fucose-1-phosphate guanylyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199328.2 | Intron | NP_001186257.2 | ||||
NM_001199329.2 | Intron | NP_001186258.2 | ||||
NM_003838.4 | Intron | NP_003829.3 |
FPGT-TNNI3K - FPGT-TNNI3K readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001112808.2 | Intron | NP_001106279.2 | ||||
NM_001199327.1 | Intron | NP_001186256.2 |
LRRIQ3 - leucine rich repeats and IQ motif containing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |