Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603811 MIM: 601891 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BANF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
BANF1 - barrier to autointegration factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143985.1 | Intron | NP_001137457.1 | ||||
NM_003860.3 | Intron | NP_003851.1 | ||||
XM_017018514.1 | Intron | XP_016874003.1 | ||||
XM_017018515.1 | Intron | XP_016874004.1 |
CST6 - cystatin E/M | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EIF1AD - eukaryotic translation initiation factor 1A domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242481.1 | Intron | NP_001229410.1 | ||||
NM_001242482.1 | Intron | NP_001229411.1 | ||||
NM_001242483.1 | Intron | NP_001229412.1 | ||||
NM_001242484.1 | Intron | NP_001229413.1 | ||||
NM_001242485.1 | Intron | NP_001229414.1 | ||||
NM_001242486.1 | Intron | NP_001229415.1 | ||||
NM_032325.3 | Intron | NP_115701.2 | ||||
XM_017018412.1 | Intron | XP_016873901.1 |
Set Membership: |
HapMap Validated |