Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTGCCTCTGGCGTGCAGTTTGTC[A/G]TCCCACCATTCTGCCCCGAGACCCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605517 MIM: 606259 MIM: 605965 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
B4GAT1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
B4GAT1 - beta-1,4-glucuronyltransferase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
BRMS1 - breast cancer metastasis suppressor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024957.1 | 693 | Silent Mutation | GAC,GAT | D,D 182 | NP_001020128.1 | |
NM_015399.3 | 693 | Silent Mutation | GAC,GAT | D,D 182 | NP_056214.1 |
LOC102724064 - uncharacterized LOC102724064 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RIN1 - Ras and Rab interactor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |