Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600526 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MARK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MARK2 - microtubule affinity regulating kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039469.2 | Intron | NP_001034558.2 | ||||
NM_001163296.1 | Intron | NP_001156768.1 | ||||
NM_001163297.1 | Intron | NP_001156769.1 | ||||
NM_004954.4 | Intron | NP_004945.4 | ||||
NM_017490.3 | Intron | NP_059672.2 | ||||
XM_011544789.1 | Intron | XP_011543091.1 | ||||
XM_011544790.1 | Intron | XP_011543092.1 | ||||
XM_011544791.1 | Intron | XP_011543093.1 | ||||
XM_011544792.1 | Intron | XP_011543094.1 | ||||
XM_011544793.1 | Intron | XP_011543095.1 | ||||
XM_011544794.1 | Intron | XP_011543096.1 | ||||
XM_011544795.1 | Intron | XP_011543097.1 | ||||
XM_011544796.1 | Intron | XP_011543098.1 | ||||
XM_011544797.1 | Intron | XP_011543099.1 | ||||
XM_011544798.1 | Intron | XP_011543100.1 | ||||
XM_017017310.1 | Intron | XP_016872799.1 | ||||
XM_017017311.1 | Intron | XP_016872800.1 |