Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
37 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607956 MIM: 615463 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MED8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MED8 - mediator complex subunit 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001653.2 | Intron | NP_001001653.1 | ||||
NM_052877.4 | Intron | NP_443109.2 | ||||
NM_201542.4 | Intron | NP_963836.2 |
SZT2 - seizure threshold 2 homolog (mouse) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015284.3 | Intron | NP_056099.3 | ||||
XM_005270686.3 | Intron | XP_005270743.1 | ||||
XM_006710501.3 | Intron | XP_006710564.1 | ||||
XM_011541106.2 | Intron | XP_011539408.1 | ||||
XM_011541107.2 | Intron | XP_011539409.1 | ||||
XM_017000819.1 | Intron | XP_016856308.1 | ||||
XM_017000820.1 | Intron | XP_016856309.1 | ||||
XM_017000821.1 | Intron | XP_016856310.1 |
Set Membership: |
HapMap |