Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGAGACCCACGACCTGGCCTGCC[A/G]TTGCCCTGAGCTGCAGCCTCGGCCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176875 MIM: 603761 MIM: 610831 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100130987 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
LOC100130987 - uncharacterized LOC100130987 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPP1CA - protein phosphatase 1 catalytic subunit alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008709.1 | 2211 | UTR 3 | NP_001008709.1 | |||
NM_002708.3 | 2211 | UTR 3 | NP_002699.1 | |||
NM_206873.1 | 2211 | UTR 3 | NP_996756.1 |
RAD9A - RAD9 checkpoint clamp component A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243224.1 | 2211 | UTR 3 | NP_001230153.1 | |||
NM_004584.2 | 2211 | UTR 3 | NP_004575.1 | |||
XM_006718652.3 | 2211 | UTR 3 | XP_006718715.1 | |||
XM_017018097.1 | 2211 | UTR 3 | XP_016873586.1 | |||
XM_017018098.1 | 2211 | UTR 3 | XP_016873587.1 |
TBC1D10C - TBC1 domain family member 10C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |