Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 603311 | ||||||||||||||||||||
Literature Links: |
CDC7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDC7 - cell division cycle 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134419.1 | Intron | NP_001127891.1 | ||||
NM_001134420.1 | Intron | NP_001127892.1 | ||||
NM_003503.3 | Intron | NP_003494.1 | ||||
XM_005271241.2 | Intron | XP_005271298.1 | ||||
XM_005271244.2 | Intron | XP_005271301.1 | ||||
XM_017002425.1 | Intron | XP_016857914.1 | ||||
XM_017002426.1 | Intron | XP_016857915.1 | ||||
XM_017002427.1 | Intron | XP_016857916.1 |