Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGCAGTATCAATGTCTCTGCTGA[C/T]TGCACTGGTCTGAAACTCCCTTTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602736 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP5G3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATP5G3 - ATP synthase, H+ transporting, mitochondrial Fo complex subunit C3 (subunit 9) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002258.4 | 373 | Missense Mutation | ATC,GTC | I,V 65 | NP_001002258.1 | |
NM_001190329.1 | 373 | Missense Mutation | ATC,GTC | I,V 65 | NP_001177258.1 | |
NM_001689.4 | 373 | Missense Mutation | ATC,GTC | I,V 65 | NP_001680.1 |