Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609557 MIM: 104614 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PREPL PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PREPL - prolyl endopeptidase-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042385.2 | 4009 | UTR 3 | NP_001035844.1 | |||
NM_001042386.2 | 4009 | UTR 3 | NP_001035845.1 | |||
NM_001171603.1 | 4009 | UTR 3 | NP_001165074.1 | |||
NM_001171606.1 | 4009 | UTR 3 | NP_001165077.1 | |||
NM_001171613.1 | 4009 | UTR 3 | NP_001165084.1 | |||
NM_001171617.1 | 4009 | UTR 3 | NP_001165088.1 | |||
NM_006036.4 | 4009 | UTR 3 | NP_006027.2 | |||
XM_011533198.1 | 4009 | UTR 3 | XP_011531500.1 | |||
XM_011533200.1 | 4009 | UTR 3 | XP_011531502.1 | |||
XM_011533202.1 | 4009 | UTR 3 | XP_011531504.1 | |||
XM_017005384.1 | 4009 | UTR 3 | XP_016860873.1 | |||
XM_017005385.1 | 4009 | UTR 3 | XP_016860874.1 |
SLC3A1 - solute carrier family 3 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000341.3 | 4009 | Intron | NP_000332.2 | |||
XM_011533047.2 | 4009 | Intron | XP_011531349.1 |
Set Membership: |
HapMap |