Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTAGTGATATAGGGAGGATAAGAC[C/T]TGTCTCTAGGATTGCTGTGATTATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603770 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPM1B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PPM1B - protein phosphatase, Mg2+/Mn2+ dependent 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033557.2 | Intron | NP_001028729.1 | ||||
NM_002706.5 | Intron | NP_002697.1 | ||||
NM_177968.3 | Intron | NP_808907.1 | ||||
NM_177969.3 | Intron | NP_808908.1 | ||||
XM_011532936.2 | Intron | XP_011531238.1 | ||||
XM_017004395.1 | Intron | XP_016859884.1 | ||||
XM_017004396.1 | Intron | XP_016859885.1 | ||||
XM_017004397.1 | Intron | XP_016859886.1 |