Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616285 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FMNL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FMNL2 - formin like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052905.3 | Intron | NP_443137.2 | ||||
XM_005246263.2 | Intron | XP_005246320.1 | ||||
XM_005246265.3 | Intron | XP_005246322.1 | ||||
XM_011510530.1 | Intron | XP_011508832.1 | ||||
XM_011510531.1 | Intron | XP_011508833.1 | ||||
XM_011510532.2 | Intron | XP_011508834.1 | ||||
XM_011510533.2 | Intron | XP_011508835.1 | ||||
XM_011510534.1 | Intron | XP_011508836.1 | ||||
XM_011510535.2 | Intron | XP_011508837.1 | ||||
XM_011510536.2 | Intron | XP_011508838.1 |