Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATGAGACTTCTTTGAAAGTGTCC[A/G]GTTTATGCTAAATATTCATGTACAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613841 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
UBN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
UBN2 - ubinuclein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173569.3 | Intron | NP_775840.3 | ||||
XM_005250249.3 | Intron | XP_005250306.2 | ||||
XM_006715916.3 | Intron | XP_006715979.2 | ||||
XM_011516002.2 | Intron | XP_011514304.1 | ||||
XM_011516003.1 | Intron | XP_011514305.1 |
Set Membership: |
HapMap Validated |