Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600834 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF165 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ZNF165 - zinc finger protein 165 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003447.3 | Intron | NP_003438.1 | ||||
XM_017011258.1 | Intron | XP_016866747.1 | ||||
XM_017011259.1 | Intron | XP_016866748.1 | ||||
XM_017011260.1 | Intron | XP_016866749.1 | ||||
XM_017011261.1 | Intron | XP_016866750.1 | ||||
XM_017011262.1 | Intron | XP_016866751.1 |
ZSCAN12P1 - zinc finger and SCAN domain containing 12 pseudogene 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
Validated |