Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGAGAGTGCTGTGTAGCTGTCCT[C/T]TTTATCCTGGTGTTTGCCACTTCGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605238 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HNMT PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
HNMT - histamine N-methyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024074.2 | 12 | Intron | NP_001019245.1 | |||
NM_001024075.1 | 12 | Intron | NP_001019246.1 | |||
NM_006895.2 | 12 | Intron | NP_008826.1 | |||
XM_011511064.2 | 12 | Intron | XP_011509366.1 | |||
XM_017003948.1 | 12 | UTR 5 | XP_016859437.1 | |||
XM_017003949.1 | 12 | Intron | XP_016859438.1 |
Set Membership: |
HapMap Validated |