Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607243 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AP4S1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AP4S1 - adaptor related protein complex 4 sigma 1 subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HECTD1 - HECT domain E3 ubiquitin protein ligase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015382.3 | 8464 | UTR 3 | NP_056197.3 | |||
XM_005267502.2 | 8464 | UTR 3 | XP_005267559.2 | |||
XM_011536619.2 | 8464 | UTR 3 | XP_011534921.1 | |||
XM_011536621.2 | 8464 | UTR 3 | XP_011534923.1 | |||
XM_011536622.1 | 8464 | UTR 3 | XP_011534924.1 | |||
XM_017021148.1 | 8464 | UTR 3 | XP_016876637.1 | |||
XM_017021149.1 | 8464 | UTR 3 | XP_016876638.1 | |||
XM_017021150.1 | 8464 | UTR 3 | XP_016876639.1 | |||
XM_017021151.1 | 8464 | UTR 3 | XP_016876640.1 | |||
XM_017021152.1 | 8464 | UTR 3 | XP_016876641.1 | |||
XM_017021153.1 | 8464 | UTR 3 | XP_016876642.1 |
Set Membership: |
HapMap |