Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611448 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BMS1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BMS1 - BMS1, ribosome biogenesis factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014753.3 | Intron | NP_055568.3 | ||||
XM_005271846.3 | Intron | XP_005271903.1 | ||||
XM_005271848.3 | Intron | XP_005271905.1 | ||||
XM_005271849.3 | Intron | XP_005271906.1 | ||||
XM_011540402.2 | Intron | XP_011538704.1 | ||||
XM_011540403.2 | Intron | XP_011538705.1 |