Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTTAGATGTTTTAACTTATTTAA[A/C]TCTGTAAACATTTCTTTTAAAAGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605882 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BRIP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BRIP1 - BRCA1 interacting protein C-terminal helicase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032043.2 | 7570 | UTR 3 | NP_114432.2 | |||
XM_011525332.2 | 7570 | Intron | XP_011523634.1 | |||
XM_011525333.2 | 7570 | Intron | XP_011523635.1 | |||
XM_011525334.2 | 7570 | Intron | XP_011523636.1 | |||
XM_011525335.2 | 7570 | Intron | XP_011523637.1 | |||
XM_011525336.2 | 7570 | Intron | XP_011523638.1 | |||
XM_011525337.2 | 7570 | Intron | XP_011523639.1 | |||
XM_011525338.2 | 7570 | Intron | XP_011523640.1 | |||
XM_011525339.2 | 7570 | Intron | XP_011523641.1 | |||
XM_011525340.2 | 7570 | Intron | XP_011523642.1 | |||
XM_011525341.2 | 7570 | Intron | XP_011523643.1 | |||
XM_017025200.1 | 7570 | Intron | XP_016880689.1 | |||
XM_017025201.1 | 7570 | Intron | XP_016880690.1 | |||
XM_017025202.1 | 7570 | Intron | XP_016880691.1 | |||
XM_017025203.1 | 7570 | Intron | XP_016880692.1 |