Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611626 MIM: 612408 | ||||||||||||||||||||
Literature Links: |
MPHOSPH8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MPHOSPH8 - M-phase phosphoprotein 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PSPC1 - paraspeckle component 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042414.2 | Intron | NP_001035879.1 | ||||
XM_006719844.3 | Intron | XP_006719907.1 | ||||
XM_011535137.1 | Intron | XP_011533439.1 | ||||
XM_011535138.2 | Intron | XP_011533440.1 | ||||
XM_011535139.1 | Intron | XP_011533441.1 | ||||
XM_011535140.1 | Intron | XP_011533442.1 | ||||
XM_011535141.2 | Intron | XP_011533443.1 | ||||
XM_011535142.2 | Intron | XP_011533444.1 | ||||
XM_017020650.1 | Intron | XP_016876139.1 |