Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
30 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616877 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM9 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TMEM9 - transmembrane protein 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288564.1 | 1490 | UTR 3 | NP_001275493.1 | |||
NM_001288565.1 | 1490 | UTR 3 | NP_001275494.1 | |||
NM_001288566.1 | 1490 | UTR 3 | NP_001275495.1 | |||
NM_001288567.1 | 1490 | UTR 3 | NP_001275496.1 | |||
NM_001288568.1 | 1490 | UTR 3 | NP_001275497.1 | |||
NM_001288569.1 | 1490 | UTR 3 | NP_001275498.1 | |||
NM_001288570.1 | 1490 | UTR 3 | NP_001275499.1 | |||
NM_001288571.1 | 1490 | UTR 3 | NP_001275500.1 | |||
NM_016456.4 | 1490 | UTR 3 | NP_057540.1 | |||
XM_011509385.1 | 1490 | UTR 3 | XP_011507687.1 | |||
XM_011509386.2 | 1490 | UTR 3 | XP_011507688.1 | |||
XM_011509387.2 | 1490 | UTR 3 | XP_011507689.1 |