Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606097 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIGN PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PIGN - phosphatidylinositol glycan anchor biosynthesis class N | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012327.5 | Intron | NP_036459.1 | ||||
NM_176787.4 | Intron | NP_789744.1 | ||||
XM_011525889.1 | Intron | XP_011524191.1 | ||||
XM_011525890.1 | Intron | XP_011524192.1 | ||||
XM_011525891.1 | Intron | XP_011524193.1 | ||||
XM_011525892.1 | Intron | XP_011524194.1 | ||||
XM_011525893.1 | Intron | XP_011524195.1 | ||||
XM_011525894.1 | Intron | XP_011524196.1 | ||||
XM_011525895.1 | Intron | XP_011524197.1 | ||||
XM_011525896.1 | Intron | XP_011524198.1 | ||||
XM_011525898.1 | Intron | XP_011524200.1 | ||||
XM_017025685.1 | Intron | XP_016881174.1 | ||||
XM_017025686.1 | Intron | XP_016881175.1 |
Set Membership: |
HapMap |