Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602246 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TOB2P1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TOB2P1 - transducer of ERBB2, 2 pseudogene 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZSCAN9 - zinc finger and SCAN domain containing 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199479.1 | 103 | UTR 5 | NP_001186408.1 | |||
NM_001199480.1 | 103 | Intron | NP_001186409.1 | |||
NM_006299.4 | 103 | UTR 5 | NP_006290.1 | |||
XM_011514875.2 | 103 | Intron | XP_011513177.1 | |||
XM_011514876.2 | 103 | Intron | XP_011513178.1 | |||
XM_011514878.2 | 103 | Intron | XP_011513180.1 | |||
XM_017011267.1 | 103 | Intron | XP_016866756.1 |
Set Membership: |
HapMap |