Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606998 MIM: 191130 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FLOT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FLOT1 - flotillin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318875.1 | 1596 | Silent Mutation | NP_001305804.1 | |||
NM_005803.3 | 1596 | Silent Mutation | NP_005794.1 | |||
XM_005248780.3 | 1596 | Silent Mutation | XP_005248837.1 | |||
XM_006714947.3 | 1596 | Silent Mutation | XP_006715010.1 | |||
XM_017010157.1 | 1596 | Silent Mutation | XP_016865646.1 | |||
XM_017010158.1 | 1596 | Silent Mutation | XP_016865647.1 |
TUBB - tubulin beta class I | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |