Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 143110 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HLA-F PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HLA-F - major histocompatibility complex, class I, F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098478.1 | Intron | NP_001091948.1 | ||||
NM_001098479.1 | Intron | NP_001091949.1 | ||||
NM_018950.2 | Intron | NP_061823.2 | ||||
XM_011514564.1 | Intron | XP_011512866.1 | ||||
XM_017010810.1 | Intron | XP_016866299.1 | ||||
XM_017010811.1 | Intron | XP_016866300.1 | ||||
XM_017010812.1 | Intron | XP_016866301.1 | ||||
XM_017010813.1 | Intron | XP_016866302.1 | ||||
XM_017010814.1 | Intron | XP_016866303.1 | ||||
XM_017010815.1 | Intron | XP_016866304.1 |
HLA-F-AS1 - HLA-F antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |