Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607719 | ||||||||||||||||||||
Literature Links: |
SYT8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SYT8 - synaptotagmin 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290332.1 | Intron | NP_001277261.1 | ||||
NM_001290333.1 | Intron | NP_001277262.1 | ||||
NM_001290334.1 | Intron | NP_001277263.1 | ||||
NM_138567.4 | Intron | NP_612634.3 | ||||
XM_011520455.1 | Intron | XP_011518757.1 | ||||
XM_011520456.2 | Intron | XP_011518758.1 | ||||
XM_011520457.2 | Intron | XP_011518759.1 | ||||
XM_017018528.1 | Intron | XP_016874017.1 | ||||
XM_017018529.1 | Intron | XP_016874018.1 |