Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 180410 MIM: 605921 | ||||||||||||||||||||
Literature Links: |
RRM1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RRM1 - ribonucleotide reductase catalytic subunit M1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033.4 | 284 | UTR 5 | NP_001024.1 | |||
NM_001318064.1 | 284 | UTR 5 | NP_001304993.1 | |||
NM_001318065.1 | 284 | Intron | NP_001304994.1 | |||
XM_011520278.1 | 284 | Intron | XP_011518580.1 |
STIM1 - stromal interaction molecule 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |