Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTATCATAAAGGTTAGGCTAAGCT[A/G]TGTAGTTAACTGACTCTGTGTTGCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
44 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607983 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GORAB PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GORAB - golgin, RAB6 interacting | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146039.1 | Intron | NP_001139511.1 | ||||
NM_001320252.1 | Intron | NP_001307181.1 | ||||
NM_152281.2 | Intron | NP_689494.2 | ||||
XM_006711628.3 | Intron | XP_006711691.1 | ||||
XM_011510149.2 | Intron | XP_011508451.1 | ||||
XM_011510150.2 | Intron | XP_011508452.1 | ||||
XM_011510151.1 | Intron | XP_011508453.1 | ||||
XM_017002807.1 | Intron | XP_016858296.1 |
LOC101928650 - uncharacterized LOC101928650 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |