Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612003 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C2orf82 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C2orf82 - chromosome 2 open reading frame 82 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_206895.1 | 310 | Intron | NP_996778.1 | |||
XM_017004080.1 | 310 | Intron | XP_016859569.1 | |||
XM_017004081.1 | 310 | UTR 5 | XP_016859570.1 | |||
XM_017004082.1 | 310 | Intron | XP_016859571.1 | |||
XM_017004083.1 | 310 | UTR 5 | XP_016859572.1 | |||
XM_017004084.1 | 310 | Intron | XP_016859573.1 |
GIGYF2 - GRB10 interacting GYF protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |